Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAG4
Entrez Gene ID
824341
Gene Name
BCL-2-associated athanogene 4
Gene Aliases
AT3G51780, ATBAG4, BCL-2-associated athanogene 4
Chromosome Location
Chr.3: 19206768 - 19208648 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115037.8 NP_190746.2 3 - 4 1087 84
GenBank mRNA AY090347.1 - 3 - 4 724 84
AY143837.1 - 3 - 4 617 84

View Full Product Details