Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK13
Entrez Gene ID
824348
Gene Name
calcium-dependent protein kinase 13
Gene Aliases
AT3G51850, calcium-dependent protein kinase 13
Chromosome Location
Chr.3: 19232358 - 19235891 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339535.1 NP_001326349.1 1 - 2 935 86
NM_001339536.1 NP_001326348.1 1 - 2 866 86
NM_115044.5 NP_190753.2 1 - 2 935 86

View Full Product Details