Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAX3
Entrez Gene ID
824349
Gene Name
cation exchanger 3
Gene Aliases
AT3G51860, ATCAX3, ATHCX1, CALCIUM/PROTON EXCHANGER CAX1-LIKE PROTEIN, CAX1-LIKE, cation exchanger 3
Chromosome Location
Chr.3: 19239305 - 19242913 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339537.1 NP_001327590.1 6 - 7 983 74
NM_115045.4 NP_190754.2 6 - 7 1090 74
GenBank mRNA AY058054.1 - 6 - 7 968 74
BT020374.1 - 6 - 7 937 74

View Full Product Details