Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCS15
Entrez Gene ID
824381
Gene Name
3-ketoacyl-CoA synthase 15
Gene Aliases
3-ketoacyl-CoA synthase 15, AT3G52160
Chromosome Location
Chr.3: 19344972 - 19346668 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115076.4 NP_190784.1 1 - 2 378 61
GenBank mRNA DQ446759.1 - 1 - 2 301 61

View Full Product Details