Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEX4
Entrez Gene ID
824383
Gene Name
dual specificity protein phosphatase (DsPTP1) family protein
Gene Aliases
AT3G52180, ATPTPKIS1, ATSEX4, DSP4, DUAL-SPECIFICITY PROTEIN PHOSPHATASE 4, DUAL-SPECIFICITY PROTEIN PHOSPHATASE 4, STARCH-EXCESS 4, T25B15.2
Chromosome Location
Chr.3: 19349600 - 19353728 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035765.2 NP_001030842.1 3 - 4 434 99
NM_001339563.1 NP_001326925.1 3 - 4 398 99
NM_115078.4 NP_566960.1 3 - 4 530 99
GenBank mRNA AJ302781.1 - 3 - 4 315 99

View Full Product Details