Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLA15
Entrez Gene ID
824402
Gene Name
FASCICLIN-like arabinogalactan protein 15 precursor
Gene Aliases
AT3G52370, FASCICLIN-like arabinogalactan protein 15 precursor
Chromosome Location
Chr.3: 19417372 - 19419913 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339573.1 NP_001325578.1 1 - 2 1427 81
NM_115097.3 NP_190805.1 1 - 2 1398 81
GenBank mRNA AK117608.1 - 1 - 2 1309 81
BT026127.1 - 1 - 2 1250 81

View Full Product Details