Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP22
Entrez Gene ID
824448
Gene Name
purple acid phosphatase 22
Gene Aliases
AT3G52820, ATPAP22, PURPLE ACID PHOSPHATASE 22, purple acid phosphatase 22
Chromosome Location
Chr.3: 19573916 - 19577141 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115142.5 NP_190850.2 5 - 6 1303 63
GenBank mRNA AF492668.1 - 5 - 6 1100 63

View Full Product Details