Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FK
Entrez Gene ID
824460
Gene Name
Ergosterol biosynthesis ERG4/ERG24 family
Gene Aliases
AT3G52940, ELL1, EXTRA-LONG-LIFESPAN 1, FACKEL, HYD2
Chromosome Location
Chr.3: 19630077 - 19633328 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115154.3 NP_566975.1 2 - 3 381 90
NM_202691.1 NP_974420.1 2 - 3 336 90
GenBank mRNA AF256535.1 - 2 - 3 289 90
AK227189.1 - 2 - 3 247 90
AY064964.1 - 2 - 3 349 90
AY133650.1 - 2 - 3 165 90

View Full Product Details