Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOS4
Entrez Gene ID
824477
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT3G53110, LOW EXPRESSION OF OSMOTICALLY RESPONSIVE GENES 4
Chromosome Location
Chr.3: 19687740 - 19690661 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115171.3 NP_190879.1 6 - 7 1360 113
GenBank mRNA BT002444.1 - 6 - 7 1311 113
BT008867.1 - 5 - 6 1229 113

View Full Product Details