Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP45
Entrez Gene ID
824491
Gene Name
receptor like protein 45
Gene Aliases
AT3G53240, AtRLP45, receptor like protein 45
Chromosome Location
Chr.3: 19733766 - 19739232 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339602.1 NP_001325565.1 2 - 3 477 125
NM_001339603.1 NP_001325566.1 2 - 3 369 125
NM_115184.5 NP_190892.3 3 - 4 668 125

View Full Product Details