Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX20
Entrez Gene ID
824539
Gene Name
cation/H+ exchanger 20
Gene Aliases
AT3G53720, ATCHX20, cation/H+ exchanger 20
Chromosome Location
Chr.3: 19905591 - 19910244 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115232.3 NP_190940.1 1 - 2 382 66
GenBank mRNA AK227229.1 - 1 - 2 270 66
BT002529.1 - 1 - 2 269 66
BT010870.1 - 1 - 2 183 66

View Full Product Details