Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX20
Entrez Gene ID
824539
Gene Name
cation/H+ exchanger 20
Gene Aliases
AT3G53720, ATCHX20, cation/H+ exchanger 20
Chromosome Location
Chr.3: 19905591 - 19910244 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339628.1 NP_001325440.1 3 - 4 2490 73
NM_115232.3 NP_190940.1 4 - 5 2370 73
GenBank mRNA AK227229.1 - 4 - 5 2258 73
BT002529.1 - 4 - 5 2257 73
BT010870.1 - 4 - 5 2171 73

View Full Product Details