Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGD3
Entrez Gene ID
824595
Gene Name
ROOT GROWTH DEFECTIVE 3
Gene Aliases
AT3G54280, ATBTAF1, BTAF1, CHA16, CHR16, ROOT GROWTH DEFECTIVE 3
Chromosome Location
Chr.3: 20091852 - 20104257 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203156.1 NP_001190085.1 14 - 15 2426 102
NM_115288.8 NP_190996.3 13 - 14 2457 102
GenBank mRNA AB440793.1 - 12 - 13 2195 102

View Full Product Details