Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPB5E
- Entrez Gene ID
-
824614
- Gene Name
- RNA polymerase II fifth largest subunit, E
- Gene Aliases
-
''RNA polymerase II fifth largest subunit, AT3G54490, E, E'', RNA polymerase II fifth largest subunit
- Chromosome Location
-
Chr.3: 20173605 - 20175462 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115306.5 |
NP_191013.1 |
3 - 4 |
638 |
76 |
View Full Product Details