Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAG1
Entrez Gene ID
824626
Gene Name
histone acetyltransferase of the GNAT family 1
Gene Aliases
AT3G54610, BGT, BIG TOP, GCN5, HAC3, HAG01, HAT1, HISTONE ACETYLTRANSFERASE 1, general control non-repressible 5, histone acetyltransferase of the GNAT family 1
Chromosome Location
Chr.3: 20213406 - 20217653 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115318.3 NP_567002.1 5 - 6 1207 115
GenBank mRNA AF338768.1 - 5 - 6 1037 115
BT002057.1 - 5 - 6 1040 115
BT008909.1 - 5 - 6 1020 115

View Full Product Details