Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BZIP25
- Entrez Gene ID
-
824627
- Gene Name
- basic leucine zipper 25
- Gene Aliases
-
ARABIDOPSIS THALIANA BASIC LEUCINE ZIPPER 25, AT3G54620, ATBZIP25, BASIC LEUCINE ZIPPER O2 HOMOLOG 4, BZO2H4, basic leucine zipper 25
- Chromosome Location
-
Chr.3: 20217854 - 20220497 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115319.4 |
NP_567003.2 |
4 - 5 |
937 |
129 |
GenBank mRNA |
AY057509.1 |
- |
4 - 5 |
832 |
129 |
View Full Product Details