Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BZIP25
Entrez Gene ID
824627
Gene Name
basic leucine zipper 25
Gene Aliases
ARABIDOPSIS THALIANA BASIC LEUCINE ZIPPER 25, AT3G54620, ATBZIP25, BASIC LEUCINE ZIPPER O2 HOMOLOG 4, BZO2H4, basic leucine zipper 25
Chromosome Location
Chr.3: 20217854 - 20220497 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125366.1 NP_001118838.1 4 - 5 1086 104
NM_115319.4 NP_567003.2 5 - 6 1069 104
GenBank mRNA AY057509.1 - 5 - 6 964 104

View Full Product Details