Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARA6
Entrez Gene ID
824649
Gene Name
Ras-related small GTP-binding family protein
Gene Aliases
ARA-6, AT3G54840, ATRAB5C, ATRABF1, AtARA6, RABF1
Chromosome Location
Chr.3: 20318412 - 20321106 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084818.1 NP_001078287.1 5 - 6 575 110
NM_001339701.1 NP_001327724.1 4 - 5 493 110
NM_115341.3 NP_567008.1 5 - 6 575 110
GenBank mRNA AB007766.2 - 5 - 6 485 110
AK316975.1 - 5 - 6 526 110
BT002860.1 - 5 - 6 480 110
BT004392.1 - 5 - 6 390 110

View Full Product Details