Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXIP1
Entrez Gene ID
824655
Gene Name
CAX interacting protein 1
Gene Aliases
AT3G54900, ATGRXCP, CAX interacting protein 1, GLUTAREDOXIN
Chromosome Location
Chr.3: 20341357 - 20342464 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115347.4 NP_191050.1 1 - 1 819 92

View Full Product Details