Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PROT2
Entrez Gene ID
824740
Gene Name
proline transporter 2
Gene Aliases
AT3G55740, ATPROT2, PROLINE TRANSPORTER 2, proline transporter 2
Chromosome Location
Chr.3: 20695433 - 20698292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339744.1 NP_001325527.1 5 - 6 1422 102
NM_115432.5 NP_191133.1 6 - 7 1300 102
NM_202715.2 NP_974444.1 6 - 7 1284 102
GenBank mRNA AY081292.1 - 6 - 7 1121 102
BT008750.1 - 6 - 7 947 102
X95738.1 - - 1098 102

View Full Product Details