Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ESK1
- Entrez Gene ID
-
824765
- Gene Name
- trichome birefringence-like protein (DUF828)
- Gene Aliases
-
AT3G55990, ESKIMO 1, TBL29, TRICHOME BIREFRINGENCE-LIKE 29
- Chromosome Location
-
Chr.3: 20780175 - 20783233 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115457.3 |
NP_191158.1 |
3 - 4 |
1192 |
67 |
View Full Product Details