Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CAN
- Entrez Gene ID
-
824783
- Gene Name
- Ca-2+ dependent nuclease
- Gene Aliases
-
AT3G56170, CAN1, Ca-2+ dependent nuclease, calcium dependent nuclease 1
- Chromosome Location
-
Chr.3: 20842145 - 20844537 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115475.5 |
NP_567036.1 |
4 - 5 |
594 |
73 |
View Full Product Details