Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPE5
Entrez Gene ID
824875
Gene Name
Eukaryotic rpb5 RNA polymerase subunit family protein
Gene Aliases
AT3G57080, B, RNA POLYMERASE II FIFTH LARGEST SUBUNIT, RPB5B
Chromosome Location
Chr.3: 21125725 - 21128075 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115567.3 NP_191267.1 2 - 3 578 95
GenBank mRNA AK118958.1 - 2 - 3 446 95
BT005410.1 - 2 - 3 425 95

View Full Product Details