Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGAL1
Entrez Gene ID
830736
Gene Name
alpha-galactosidase 1
Gene Aliases
AT5G08380, AtAGAL1, F8L15.110, F8L15_110, alpha-galactosidase 1
Chromosome Location
Chr.5: 2694669 - 2697781 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342998.1 NP_001332077.1 2 - 3 336 79
NM_120922.5 NP_196455.1 2 - 3 393 79

View Full Product Details