Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KUP7
Entrez Gene ID
830799
Gene Name
K+ uptake permease 7
Gene Aliases
AT5G09400, K+ uptake permease 7, T5E8.200, T5E8_200
Chromosome Location
Chr.5: 2916151 - 2920823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120977.3 NP_568213.2 7 - 8 1477 78

View Full Product Details