Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TOC64-V
- Entrez Gene ID
-
830801
- Gene Name
- translocon at the outer membrane of chloroplasts 64-V
- Gene Aliases
-
ARABIDOPSIS THALIANA TRANSLOCON AT THE OUTER MEMBRANE OF CHLOROPLASTS 64-V, AT5G09420, ATTOC64-V, AtmtOM64, MTOM64, OM64, T5E8.220, T5E8_220, outer membrane 64, translocon at the outer membrane of chloroplasts 64-V
- Chromosome Location
-
Chr.5: 2928173 - 2932021 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120979.3 |
NP_196504.2 |
6 - 7 |
1115 |
91 |
View Full Product Details