Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCME
Entrez Gene ID
831443
Gene Name
prenylcysteine methylesterase
Gene Aliases
AT5G15860, ATPCME, F14F8.240, F14F8_240, ICME, Isoprenylcysteine methylesterase, prenylcysteine methylesterase
Chromosome Location
Chr.5: 5178157 - 5181761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121591.5 NP_197090.2 5 - 6 1081 97
NM_203057.1 NP_974786.1 5 - 6 610 97
GenBank mRNA AY045829.1 - 5 - 6 702 97
AY091367.1 - 5 - 6 583 97

View Full Product Details