Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TT16
Entrez Gene ID
832390
Gene Name
K-box region and MADS-box transcription factor family protein
Gene Aliases
ABS, AGAMOUS-like 32, AGL32, ARABIDOPSIS BSISTER, AT5G23260, MKD15.12, MKD15_12, TRANSPARENT TESTA16
Chromosome Location
Chr.5: 7835965 - 7838555 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343796.1 NP_001318626.1 6 - 7 863 96
NM_001343797.1 NP_001330404.1 6 - 7 878 96
NM_203094.2 NP_974823.1 6 - 7 727 96
GenBank mRNA AJ318098.1 - 6 - 7 596 96

View Full Product Details