Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARP9
Entrez Gene ID
834370
Gene Name
actin-related protein 9
Gene Aliases
AT5G43500, ATARP9, MWF20.22, MWF20_22, actin-related protein 9
Chromosome Location
Chr.5: 17469490 - 17473619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344487.1 NP_001331886.1 10 - 11 1454 93
NM_001344488.1 NP_001331887.1 10 - 11 1296 93
NM_123716.3 NP_199163.1 10 - 11 1408 93
NM_203147.2 NP_974876.1 10 - 11 1454 93
GenBank mRNA AY057568.1 - 10 - 11 1223 93

View Full Product Details