Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPOX
Entrez Gene ID
835061
Gene Name
pyridoxin (pyrodoxamine) 5'-phosphate oxidase
Gene Aliases
AT5G49970, ATPPOX, HOMOLOG OF YEAST PYRIDOXINE AUXOTROPHY 3, K9P8.11, K9P8_11, PDX3, pyridoxin (pyrodoxamine) 5'-phosphate oxidase
Chromosome Location
Chr.5: 20329103 - 20333172 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124376.4 NP_568717.2 4 - 5 727 106
NM_203189.2 NP_974918.1 4 - 5 628 106

View Full Product Details