Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGP1
Entrez Gene ID
835574
Gene Name
Ras-related small GTP-binding family protein
Gene Aliases
AT5G54840, ATSGP1, MBG8.10, MBG8_10
Chromosome Location
Chr.5: 22276343 - 22278423 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345101.1 NP_001331825.1 3 - 4 673 147
NM_124865.4 NP_200295.1 3 - 4 673 147
NM_203204.3 NP_974933.1 3 - 4 673 147
GenBank mRNA AJ238846.1 - 3 - 4 593 147
BT030087.1 - 3 - 4 578 147

View Full Product Details