Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPF3
Entrez Gene ID
840295
Gene Name
Smg-4/UPF3 family protein
Gene Aliases
AT1G33980, ATUPF3, F12G12.20, F12G12_20
Chromosome Location
Chr.1: 12351566 - 12355173 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123934.1 NP_001117406.1 11 - 12 1545 117
NM_103120.4 NP_174660.2 11 - 12 1566 117
GenBank mRNA AY128383.1 - 11 - 12 1526 117
BT000112.1 - 11 - 12 1413 117

View Full Product Details