Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG10
Entrez Gene ID
840369
Gene Name
homeodomain GLABROUS 10
Gene Aliases
AT1G34650, F21H2.11, F21H2_11, homeodomain GLABROUS 10
Chromosome Location
Chr.1: 12692995 - 12697834 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103188.3 NP_174724.1 6 - 7 1234 75

View Full Product Details