Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SHM7
- Entrez Gene ID
-
840543
- Gene Name
- serine hydroxymethyltransferase 7
- Gene Aliases
-
AT1G36370, F7F23.9, F7F23_9, serine hydroxymethyltransferase 7
- Chromosome Location
-
Chr.1: 13696042 - 13698739 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103323.3 |
NP_564473.1 |
3 - 4 |
1732 |
124 |
View Full Product Details