Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKS6
Entrez Gene ID
840792
Gene Name
SKU5-similar 6
Gene Aliases
AT1G41830, F5A13.5, F5A13_5, SKU5 SIMILAR 6, SKU5-similar 6
Chromosome Location
Chr.1: 15603568 - 15607966 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103408.3 NP_564479.1 1 - 2 308 60

View Full Product Details