Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MLO9
- Entrez Gene ID
-
840861
- Gene Name
- Seven transmembrane MLO family protein
- Gene Aliases
-
ARABIDOPSIS THALIANA MILDEW RESISTANCE LOCUS O 9, AT1G42560, ATMLO9, MILDEW RESISTANCE LOCUS O 9, T8D8.5, T8D8_5
- Chromosome Location
-
Chr.1: 15980135 - 15983571 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103440.4 |
NP_174980.3 |
14 - 15 |
1530 |
118 |
View Full Product Details