Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCV2
Entrez Gene ID
840912
Gene Name
like COV 2
Gene Aliases
AT1G43130, F1I21.3, F1I21_3, like COV 2
Chromosome Location
Chr.1: 16228189 - 16231254 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333208.1 NP_001321430.1 5 - 6 725 108
NM_001333209.1 NP_001321431.1 4 - 5 658 108
NM_103466.5 NP_564483.1 4 - 5 545 108
GenBank mRNA AK176692.1 - 4 - 5 524 108
AK176760.1 - 4 - 5 524 108
BT033158.1 - 4 - 5 449 108

View Full Product Details