Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLP5
Entrez Gene ID
840948
Gene Name
tubby like protein 5
Gene Aliases
AT1G43640, AtTLP5, T10P12.9, T10P12_9, tubby like protein 5
Chromosome Location
Chr.1: 16439367 - 16441941 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333218.1 NP_001320745.1 4 - 5 694 97
NM_103489.3 NP_564485.1 4 - 5 788 97
GenBank mRNA AF424565.1 - 4 - 5 690 97
AY046921.1 - 3 - 4 571 97
AY072154.1 - 4 - 5 690 97
AY122972.1 - 3 - 4 571 97

View Full Product Details