Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- scpl44
- Entrez Gene ID
-
840974
- Gene Name
- serine carboxypeptidase-like 44
- Gene Aliases
-
AT1G43780, F28H19.5, F28H19_5, serine carboxypeptidase-like 44
- Chromosome Location
-
Chr.1: 16563811 - 16567399 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103506.1 |
NP_175046.1 |
3 - 4 |
467 |
96 |
View Full Product Details