Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH3H1
Entrez Gene ID
841020
Gene Name
aldehyde dehydrogenase 3H1
Gene Aliases
ALDH4, AT1G44170, T7O23.15, T7O23_15, aldehyde dehydrogenase 3H1, aldehyde dehydrogenase 4
Chromosome Location
Chr.1: 16796400 - 16800321 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084207.1 NP_001077676.1 4 - 5 495 66
NM_103541.3 NP_175081.1 5 - 6 647 66
NM_179439.4 NP_849770.1 4 - 5 859 66
GenBank mRNA AY072122.1 - 4 - 5 761 66

View Full Product Details