Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hemb2
Entrez Gene ID
841025
Gene Name
Aldolase superfamily protein
Gene Aliases
AT1G44318, T18F15.10, T18F15_10
Chromosome Location
Chr.1: 16831016 - 16834011 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103545.2 NP_175085.1 3 - 4 426 60

View Full Product Details