Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HSFB4
- Entrez Gene ID
-
841110
- Gene Name
- heat shock transcription factor B4
- Gene Aliases
-
AT-HSFB4, AT1G46264, F2G19.8, F2G19_8, SCHIZORIZA, SCZ, heat shock transcription factor B4, heat shock transcription factor B4
- Chromosome Location
-
Chr.1: 17224720 - 17226541 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103602.3 |
NP_175142.1 |
1 - 2 |
467 |
117 |
GenBank mRNA |
AB493499.1 |
- |
1 - 2 |
283 |
117 |
View Full Product Details