Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HXK3
Entrez Gene ID
841200
Gene Name
hexokinase 3
Gene Aliases
AT1G47840, T2E6.5, T2E6_5, hexokinase 3
Chromosome Location
Chr.1: 17616051 - 17619011 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333285.1 NP_001321728.1 1 - 2 409 109
NM_103682.4 NP_175220.2 1 - 2 429 109
GenBank mRNA BT030472.1 - 1 - 2 277 109

View Full Product Details