Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GA2OX4
Entrez Gene ID
841217
Gene Name
gibberellin 2-oxidase 4
Gene Aliases
AT1G47990, ATGA2OX4, Arabidopsis thaliana gibberellin 2-oxidase 4, T2J15.10, T2J15_10, gibberellin 2-oxidase 4
Chromosome Location
Chr.1: 17698598 - 17701041 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103695.2 NP_175233.1 2 - 3 766 86
GenBank mRNA AY859740.1 - 2 - 3 709 86

View Full Product Details