Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PER1
- Entrez Gene ID
-
841231
- Gene Name
- 1-cysteine peroxiredoxin 1
- Gene Aliases
-
1-CYS PEROXIREDOXIN, 1-cysteine peroxiredoxin 1, AT1G48130, ATPER1, F21D18.15, F21D18_15
- Chromosome Location
-
Chr.1: 17780429 - 17781727 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103709.4 |
NP_175247.1 |
1 - 2 |
298 |
147 |
View Full Product Details