Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLN1;5
Entrez Gene ID
841268
Gene Name
glutamine synthetase 1;5
Gene Aliases
AT1G48470, T1N15.8, T1N15_8, glutamine synthetase 1;5
Chromosome Location
Chr.1: 17913711 - 17916101 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103743.3 NP_175280.1 7 - 8 737 70
GenBank mRNA AK118005.1 - 7 - 8 737 70
BT006245.1 - 7 - 8 664 70

View Full Product Details