Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TT12
Entrez Gene ID
825072
Gene Name
MATE efflux family protein
Gene Aliases
A. THALIANA TRANSPARENT TESTA, AT3G59030, ATTT12, TRANSPARENT TESTA 12
Chromosome Location
Chr.3: 21819040 - 21821440 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115765.4 NP_191462.1 6 - 7 1448 129
GenBank mRNA AJ294464.1 - 6 - 7 1400 129
AY088768.1 - 6 - 7 1417 129

View Full Product Details