Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSL11
Entrez Gene ID
825079
Gene Name
glucan synthase-like 11
Gene Aliases
AT3G59100, ATGSL11, glucan synthase-like 11, gsl11
Chromosome Location
Chr.3: 21842964 - 21854117 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339950.1 NP_001326589.1 35 - 36 4931 75
NM_115772.4 NP_191469.3 41 - 42 5959 75

View Full Product Details