Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HLECRK
- Entrez Gene ID
-
825139
- Gene Name
- lectin-receptor kinase
- Gene Aliases
-
AT3G59700, ATHLECRK, L-type lectin receptor kinase V.5, LECRK1, LECTIN-RECEPTOR KINASE 1, LEGUME-LIKE LECTIN RECEPTOR KINASE-V.5, lectin-receptor kinase
- Chromosome Location
-
Chr.3: 22052121 - 22054394 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115832.3 |
NP_191529.1 |
1 - 1 |
2093 |
134 |
GenBank mRNA |
X91630.1 |
- |
- |
2081 |
134 |
View Full Product Details