Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK10
Entrez Gene ID
825148
Gene Name
MAP kinase 10
Gene Aliases
AT3G59790, ATMPK10, MAP kinase 10
Chromosome Location
Chr.3: 22092400 - 22094403 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115841.2 NP_191538.1 5 - 6 1061 90

View Full Product Details